Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mc3r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025270.3 | NP_001020441.3 | 1 | 569 | ||
GenBank mRNA | AY671938.1 | - | 1 | 575 | ||
X70667.1 | - | 1 | 767 |
Set Membership: |
G-protein coupled receptor |