Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031574.1 | NP_113762.1 | 3, 4 | 284 | ||
GenBank mRNA | D86045.1 | - | 3, 4 | 284 |