Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053304.1 | NP_445756.1 | 51 | 4354 | ||
GenBank mRNA | BC133728.1 | - | 51 | 4338 | 7125649 | |
M12199.1 | - | 1 | 40 | |||
Z78279.1 | - | 51 | 4243 |