Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACTA1 SIRNA Assays ›
Gene Symbol:
Acta1
Gene Name:
actin, alpha 1, skeletal muscle
Gene Aliases:
-
Chromosome Location:
Chr.19: 56674072 - 56677084 on Build Rnor_6.0
UniGene ID:
Rn.82732
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s131418
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Acta1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019212.2
NP_062085.1
3
458
GenBank mRNA
BC061974.1
-
3
458
5623909
FQ214583.1
-
3
467
FQ214598.1
-
3
467
FQ214600.1
-
3
467
FQ214603.1
-
3
467
FQ214610.1
-
1
12
FQ214623.1
-
3
467
FQ214640.1
-
3
467
FQ214659.1
-
3
469
FQ214676.1
-
3
467
FQ214680.1
-
3
467
FQ214697.1
-
3
467
FQ214734.1
-
3
467
FQ214743.1
-
3
467
FQ214756.1
-
3
467
FQ214761.1
-
3
466
FQ214779.1
-
3
467
FQ214785.1
-
3
465
FQ214786.1
-
3
467
FQ214807.1
-
3
467
FQ214814.1
-
3
467
FQ214822.1
-
3
467
FQ214831.1
-
3
467
FQ214832.1
-
3
467
FQ214839.1
-
3
470
FQ214841.1
-
3
467
FQ214842.1
-
3
467
FQ214857.1
-
3
471
FQ214858.1
-
3
467
FQ214859.1
-
3
467
FQ214860.1
-
3
467
FQ214862.1
-
3
467
FQ214866.1
-
3
467
FQ214883.1
-
3
466
FQ214904.1
-
3
467
FQ214905.1
-
3
466
FQ214916.1
-
3
467
FQ214936.1
-
3
467
FQ214948.1
-
3
467
FQ214949.1
-
3
467
FQ214957.1
-
3
467
FQ214969.1
-
3
467
FQ214975.1
-
3
469
FQ214976.1
-
3
467
FQ214978.1
-
3
467
FQ214987.1
-
3
467
FQ214990.1
-
3
467
FQ214993.1
-
3
467
FQ215012.1
-
3
467
FQ215025.1
-
3
467
FQ215038.1
-
3
467
FQ215044.1
-
3
467
FQ215058.1
-
3
467
FQ215089.1
-
3
467
FQ215112.1
-
3
467
FQ215128.1
-
3
469
FQ215149.1
-
3
467
FQ215153.1
-
3
467
FQ215158.1
-
3
465
FQ215173.1
-
3
467
FQ215212.1
-
3
467
FQ215215.1
-
3
467
FQ215225.1
-
3
467
FQ215241.1
-
3
467
FQ215249.1
-
3
468
FQ215253.1
-
3
467
FQ215256.1
-
3
467
FQ215275.1
-
3
467
FQ215277.1
-
3
468
FQ215280.1
-
3
467
FQ215282.1
-
3
467
FQ215298.1
-
3
469
FQ215324.1
-
3
467
FQ215333.1
-
3
467
FQ215342.1
-
3
467
FQ215346.1
-
3
467
FQ215366.1
-
3
467
FQ215380.1
-
3
467
FQ215381.1
-
3
467
FQ215393.1
-
3
467
FQ215396.1
-
3
467
FQ215398.1
-
3
467
FQ215402.1
-
3
467
FQ215403.1
-
3
467
FQ215405.1
-
3
467
FQ215414.1
-
3
469
FQ215431.1
-
3
467
FQ215433.1
-
3
466
FQ215434.1
-
3
469
FQ215438.1
-
3
467
FQ215446.1
-
3
467
FQ215449.1
-
3
469
FQ215454.1
-
3
467
FQ215464.1
-
3
467
FQ215465.1
-
3
467
FQ215468.1
-
3
467
FQ215470.1
-
3
467
FQ215474.1
-
3
467
FQ215483.1
-
3
467
FQ215488.1
-
3
467
FQ215518.1
-
3
476
FQ215523.1
-
3
467
FQ215537.1
-
3
469
FQ215554.1
-
3
467
FQ215567.1
-
3
467
FQ215572.1
-
3
467
FQ215593.1
-
3
467
FQ215618.1
-
3
467
FQ215628.1
-
3
467
FQ215631.1
-
1
81
FQ215655.1
-
3
465
FQ215659.1
-
3
466
FQ215673.1
-
3
467
FQ215674.1
-
3
467
FQ215690.1
-
3
467
FQ215692.1
-
3
467
FQ215785.1
-
3
467
FQ215790.1
-
3
469
FQ215799.1
-
3
467
FQ215809.1
-
3
467
FQ215813.1
-
3
467
FQ215835.1
-
3
467
FQ215836.1
-
3
467
FQ215839.1
-
3
467
FQ215840.1
-
3
467
FQ215841.1
-
3
467
FQ215851.1
-
3
467
FQ215852.1
-
3
465
FQ215864.1
-
3
468
FQ215866.1
-
3
469
FQ215888.1
-
3
466
FQ215893.1
-
3
468
FQ215896.1
-
3
467
FQ215907.1
-
3
469
FQ215924.1
-
1
159
FQ215950.1
-
3
467
FQ215964.1
-
3
467
FQ215972.1
-
3
467
FQ216016.1
-
3
467
FQ216035.1
-
3
467
FQ216043.1
-
3
467
FQ216050.1
-
3
467
FQ216066.1
-
3
465
FQ216138.1
-
3
467
FQ216139.1
-
3
467
FQ216168.1
-
3
465
FQ216196.1
-
3
467
FQ216224.1
-
3
467
FQ216246.1
-
3
467
FQ216276.1
-
3
467
FQ216278.1
-
3
469
FQ216287.1
-
3
465
FQ216291.1
-
3
467
FQ216293.1
-
3
467
FQ216305.1
-
3
467
FQ216329.1
-
3
467
FQ216333.1
-
3
467
FQ216350.1
-
3
467
FQ216352.1
-
3
467
FQ216360.1
-
3
467
FQ216369.1
-
3
469
FQ216403.1
-
3
467
FQ216430.1
-
3
467
FQ216509.1
-
3
465
FQ216586.1
-
3
467
FQ216603.1
-
3
467
FQ216606.1
-
3
467
FQ216607.1
-
3
463
FQ216610.1
-
3
465
FQ216620.1
-
3
467
FQ216635.1
-
3
465
FQ216639.1
-
3
467
FQ216644.1
-
3
467
FQ216664.1
-
3
465
FQ216675.1
-
3
467
FQ216676.1
-
3
467
FQ216692.1
-
3
467
FQ216703.1
-
3
467
FQ216713.1
-
3
467
FQ216721.1
-
3
467
FQ216723.1
-
3
467
FQ216725.1
-
3
463
FQ216739.1
-
3
467
FQ216762.1
-
3
465
FQ216767.1
-
3
467
FQ216776.1
-
3
467
FQ216777.1
-
3
467
FQ216793.1
-
3
467
FQ216801.1
-
3
467
FQ216814.1
-
3
461
FQ216819.1
-
3
465
FQ216916.1
-
3
467
FQ216918.1
-
1
110
FQ216961.1
-
3
467
FQ216994.1
-
3
467
FQ217061.1
-
3
469
FQ217073.1
-
3
470
FQ217091.1
-
3
466
FQ217114.1
-
3
467
FQ217122.1
-
3
467
FQ217167.1
-
3
467
FQ217173.1
-
3
467
FQ217188.1
-
3
467
FQ217194.1
-
3
467
FQ217273.1
-
3
466
FQ217280.1
-
3
587
FQ217327.1
-
3
467
FQ217329.1
-
3
467
FQ217347.1
-
3
467
FQ217353.1
-
3
467
FQ217367.1
-
3
467
FQ217397.1
-
3
467
FQ217441.1
-
3
467
FQ217443.1
-
3
469
FQ217475.1
-
3
467
FQ217525.1
-
3
467
FQ217535.1
-
3
467
FQ217572.1
-
3
467
FQ217593.1
-
3
467
FQ217602.1
-
3
467
FQ217615.1
-
3
467
FQ217629.1
-
3
467
FQ217641.1
-
3
464
FQ217762.1
-
3
467
FQ217826.1
-
3
467
FQ217884.1
-
3
465
FQ217893.1
-
3
467
FQ217905.1
-
3
467
FQ217908.1
-
3
467
FQ217919.1
-
3
467
FQ217937.1
-
3
467
FQ217969.1
-
3
469
FQ217990.1
-
3
467
FQ218019.1
-
3
467
FQ218034.1
-
3
467
FQ223563.1
-
3
467
FQ223681.1
-
3
469
FQ223686.1
-
3
467
FQ223702.1
-
3
467
FQ223727.1
-
1
165
FQ223898.1
-
3
467
FQ223914.1
-
3
467
FQ223926.1
-
3
467
FQ223929.1
-
3
467
FQ224009.1
-
3
467
FQ224082.1
-
3
467
FQ224141.1
-
3
468
FQ224167.1
-
3
467
FQ224359.1
-
3
467
FQ224377.1
-
3
467
FQ224419.1
-
3
467
FQ224691.1
-
3
465
FQ224777.1
-
3
467
FQ224840.1
-
3
465
FQ224842.1
-
3
467
FQ224862.1
-
3
467
FQ224869.1
-
3
467
FQ224886.1
-
3
467
FQ224890.1
-
3
465
FQ224902.1
-
3
467
FQ224929.1
-
3
465
FQ224936.1
-
3
467
FQ224942.1
-
3
469
FQ224948.1
-
3
469
FQ224962.1
-
3
467
FQ224971.1
-
3
467
Back To Top
More Information
Panther Classification:
Molecular Function -
actin and actin related protein
actin or actin-binding cytoskeletal protein
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
response to mechanical stimulus
response to extracellular stimulus
response to lithium ion
cell growth
skeletal muscle thin filament assembly
skeletal muscle fiber adaptation
response to steroid hormone
skeletal muscle fiber development
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline