Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rab7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023950.3 | NP_076440.1 | 3 | 268 | ||
XM_006236849.2 | XP_006236911.1 | 3 | 219 | |||
GenBank mRNA | AB158427.1 | - | 2 | 100 | ||
AB158428.1 | - | 2 | 100 | |||
AF286535.1 | - | 3 | 264 | |||
BC072470.1 | - | 3 | 268 | 7098657 | ||
X12535.1 | - | 3 | 141 |