Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111114.1 | NP_001104584.1 | 2 | 382 | ||
NM_001111117.1 | NP_001104587.1 | 2 | 382 | |||
NM_017241.2 | NP_058937.1 | 2 | 382 | |||
XM_006248053.2 | XP_006248115.1 | 2 | 766 | |||
XM_006248054.2 | XP_006248116.1 | 2 | 766 | |||
GenBank mRNA | M83560.1 | - | 2 | 384 | ||
M83561.1 | - | 2 | 384 | |||
Z11712.1 | - | 2 | 228 | |||
Z11713.1 | - | 2 | 228 | |||
Z11714.1 | - | 2 | 228 |