Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017254.1 | NP_058950.1 | 2 | 519 | ||
GenBank mRNA | M30705.1 | - | 2 | 519 | ||
M64867.1 | - | 925 | ||||
X13971.1 | - | 1062 |
Set Membership: |
![]() |