Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017264.3 | NP_058960.2 | 6 | 473 | ||
GenBank mRNA | BC060574.1 | - | 6 | 477 | 6890794 | |
D45249.1 | - | 6 | 397 | |||
FQ216423.1 | - | 6 | 474 | |||
FQ219304.1 | - | 6 | 476 | |||
FQ220175.1 | - | 6 | 422 | |||
FQ224986.1 | - | 6 | 420 | |||
FQ228562.1 | - | 428 | ||||
FQ230622.1 | - | 6 | 420 | |||
FQ233032.1 | - | 6 | 426 | |||
FQ233635.1 | - | 6 | 426 | |||
FQ233859.1 | - | 6 | 422 |