Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017264.3 | NP_058960.2 | 7 | 538 | ||
GenBank mRNA | BC060574.1 | - | 7 | 542 | 6890794 | |
D45249.1 | - | 7 | 462 | |||
FQ216423.1 | - | 7 | 539 | |||
FQ219304.1 | - | 7 | 541 | |||
FQ220175.1 | - | 7 | 487 | |||
FQ224986.1 | - | 7 | 485 | |||
FQ226855.1 | - | 7 | 491 | |||
FQ228562.1 | - | 493 | ||||
FQ230622.1 | - | 7 | 485 | |||
FQ233032.1 | - | 7 | 491 | |||
FQ233635.1 | - | 7 | 491 | |||
FQ233859.1 | - | 7 | 487 |