Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017275.1 | NP_058971.1 | 5 | 508 | ||
XM_006229572.2 | XP_006229634.1 | 5 | 340 | |||
XM_006229573.2 | XP_006229635.1 | 5 | 364 | |||
XM_006229574.2 | XP_006229636.1 | 5 | 314 | |||
XM_006229575.2 | XP_006229637.1 | 5 | 318 | |||
XM_008773628.1 | XP_008771850.1 | 5 | 433 | |||
XM_008773629.1 | XP_008771851.1 | 4 | 311 | |||
GenBank mRNA | AB004267.1 | - | 5 | 508 | ||
D86556.1 | - | 5 | 359 |