Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301069.1 | NP_001287998.1 | 7 | 917 | ||
NM_003089.5 | NP_003080.2 | 7 | 917 | |||
XM_011527240.2 | XP_011525542.2 | 2 | 136 | |||
GenBank mRNA | AK096783.1 | - | 7 | 606 | ||
AL560281.3 | - | 7 | 605 | |||
BC001315.1 | - | 7 | 590 | 3454479 | ||
M22636.1 | - | 7 | 581 | |||
X06812.1 | - | 5 | 265 | |||
X06815.1 | - | 7 | 561 | |||
X84841.1 | - | 6 | 410 |