Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017304.2 | NP_059000.1 | 3, 4 | 594 | ||
XM_008764342.1 | XP_008762564.1 | 3, 4 | 360 | |||
XM_008764343.1 | XP_008762565.1 | 3, 4 | 613 | |||
GenBank mRNA | X76724.1 | - | 3, 4 | 704 |