Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kcnab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017304.2 | NP_059000.1 | 6 | 728 | ||
XM_008764342.1 | XP_008762564.1 | 6 | 494 | |||
XM_008764343.1 | XP_008762565.1 | 6 | 747 | |||
XM_008764344.1 | XP_008762566.1 | 5 | 451 | |||
XM_008764345.1 | XP_008762567.1 | 4 | 336 | |||
GenBank mRNA | X76724.1 | - | 6 | 838 |