Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017308.1 | NP_059004.1 | 3 | 287 | ||
XM_006232858.2 | XP_006232920.1 | 6 | 699 | |||
XM_006232861.2 | XP_006232923.1 | 4 | 561 | |||
XM_006232869.2 | XP_006232931.1 | 2 | 208 | |||
XM_008761286.1 | XP_008759508.1 | 4 | 3965 | |||
XM_008761287.1 | XP_008759509.1 | 4 | 3312 | |||
XM_008761288.1 | XP_008759510.1 | 4 | 2238 | |||
XM_008761289.1 | XP_008759511.1 | 4 | 3817 | |||
XM_008761290.1 | XP_008759512.1 | 4 | 3064 | |||
XM_008761291.1 | XP_008759513.1 | 4 | 3395 | |||
XM_008761292.1 | XP_008759514.1 | 4 | 2494 | |||
XM_008761293.1 | XP_008759515.1 | 5 | 651 | |||
XM_008761294.1 | XP_008759516.1 | 3 | 4123 | |||
GenBank mRNA | AB000817.2 | - | 3 | 303 | ||
AB014074.1 | - | 3 | 303 | |||
AF363971.1 | - | 3 | 287 |