Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017308.1 | NP_059004.1 | 7 | 669 | ||
XM_006232858.2 | XP_006232920.1 | 10 | 1081 | |||
XM_006232861.2 | XP_006232923.1 | 8 | 943 | |||
XM_006232869.2 | XP_006232931.1 | 6 | 590 | |||
XM_008761286.1 | XP_008759508.1 | 8 | 4347 | |||
XM_008761287.1 | XP_008759509.1 | 8 | 3694 | |||
XM_008761288.1 | XP_008759510.1 | 8 | 2620 | |||
XM_008761289.1 | XP_008759511.1 | 8 | 4199 | |||
XM_008761290.1 | XP_008759512.1 | 8 | 3446 | |||
XM_008761291.1 | XP_008759513.1 | 8 | 3777 | |||
XM_008761292.1 | XP_008759514.1 | 8 | 2876 | |||
XM_008761293.1 | XP_008759515.1 | 9 | 1033 | |||
XM_008761294.1 | XP_008759516.1 | 7 | 4505 | |||
GenBank mRNA | AB000817.2 | - | 7 | 685 | ||
AB014074.1 | - | 7 | 685 |