Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003091.3 | NP_003082.1 | 3 | 421 | ||
NM_198216.1 | NP_937859.1 | 3 | 421 | |||
GenBank mRNA | AK301034.1 | - | 4 | 477 | ||
AK301206.1 | - | 3 | 355 | |||
AK312620.1 | - | 3 | 356 | |||
AY742712.1 | - | 2 | 258 | |||
BC080516.1 | - | 3 | 339 | 2822685 | ||
BM559678.1 | - | 421 | ||||
CR456969.1 | - | 2 | 258 | |||
J04564.1 | - | 3 | 384 | |||
M34081.1 | - | 2 | 237 | |||
M34082.1 | - | 2 | 237 | |||
X15893.1 | - | 2 | 258 | |||
X17567.1 | - | 3 | 312 | |||
X17568.1 | - | 3 | 297 |