Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019275.2 | NP_062148.1 | 5 | 770 | ||
GenBank mRNA | AB010954.1 | - | 5 | 770 | ||
AF056002.1 | - | 842 |