Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ednrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017333.1 | NP_059029.1 | 4 | 1027 | ||
XM_006252431.2 | XP_006252493.1 | 4 | 1308 | |||
GenBank mRNA | S65355.1 | - | 4 | 1027 | ||
X57764.1 | - | 3 | 976 |
Set Membership: |
![]() |