Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019326.1 | NP_062199.1 | 1 | 1097 | ||
GenBank mRNA | D82868.1 | - | 1 | 1097 |