Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019328.3 | NP_062201.2 | 2 | 377 | ||
XM_006234201.2 | XP_006234263.1 | 3 | 595 | |||
XM_008761861.1 | XP_008760083.1 | 2 | 511 | |||
GenBank mRNA | DQ092629.1 | - | 2 | 308 | ||
DQ092630.1 | - | 2 | 144 | |||
DQ092631.1 | - | 2 | 322 | |||
DQ092632.1 | - | 2 | 312 | |||
DQ092633.1 | - | 2 | 144 | |||
DQ092634.1 | - | 2 | 144 | |||
FQ222109.1 | - | 3 | 616 | |||
L08595.1 | - | 2 | 377 | |||
U01146.1 | - | 2 | 308 | |||
U72345.1 | - | 3 | 601 |