Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019352.2 | NP_062225.1 | 6 | 587 | ||
GenBank mRNA | AB006451.1 | - | 6 | 587 | ||
BC083559.1 | - | 6 | 594 | 7135081 | ||
FQ216304.1 | - | 6 | 591 | |||
FQ229692.1 | - | 6 | 592 | |||
FQ229803.1 | - | 6 | 593 |
Gene Symbol: LOC100362432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729199.4 | XP_002729245.1 | 1 | 700 |