Gene Symbol: SOAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252511.1 | NP_001239440.1 | 7 | 1010 | ||
NM_001252512.1 | NP_001239441.1 | 7 | 943 | |||
NM_003101.5 | NP_003092.4 | 8 | 1069 | |||
NR_045530.1 | 6 | 988 | ||||
XM_011509911.1 | XP_011508213.1 | 8 | 1257 | |||
XM_011509912.1 | XP_011508214.1 | 8 | 1064 | |||
GenBank mRNA | AK026611.1 | - | 8 | 981 | ||
AK290659.1 | - | 7 | 927 | |||
AK294047.1 | - | 6 | 905 | |||
AK300551.1 | - | 7 | 867 | |||
AK310920.1 | - | 6 | 861 | |||
AK316401.1 | - | 7 | 772 | |||
AL833625.1 | - | 7 | 915 | |||
BC028940.1 | - | 8 | 901 | 4991365 | ||
L21934.2 | - | 2234 |