Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LOC56764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020089.2 | NP_064474.1 | 1 | 727 | ||
GenBank mRNA | AF154849.1 | - | 1 | 701 | ||
BC083558.1 | - | 1 | 727 | 7134067 |