Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005632.2 | NP_005623.1 | 7 | 2380 | ||
XM_011522620.2 | XP_011520922.1 | 8 | 5679 | |||
XM_011522621.2 | XP_011520923.1 | 8 | 6607 | |||
XM_011522622.1 | XP_011520924.1 | 6 | 5311 | |||
XM_011522623.2 | XP_011520925.1 | 9 | 3382 | |||
XM_011522624.2 | XP_011520926.1 | 8 | 2720 | |||
XM_011522625.2 | XP_011520927.1 | 7 | 2606 | |||
XM_011522626.2 | XP_011520928.1 | 8 | 6171 | |||
XM_011522627.2 | XP_011520929.1 | 8 | 6176 | |||
XM_011522628.2 | XP_011520930.1 | 8 | 3240 | |||
XM_011522629.2 | XP_011520931.1 | 8 | 3103 | |||
XM_011522630.2 | XP_011520932.1 | 7 | 2441 | |||
XM_011522631.2 | XP_011520933.1 | 6 | 2327 | |||
XM_011522632.2 | XP_011520934.1 | 6 | 2758 | |||
XM_017023596.1 | XP_016879085.1 | 9 | 3519 | |||
GenBank mRNA | BC021854.1 | - | 3 | 447 | ||
U85647.1 | - | 7 | 2380 |