Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYL1 SIRNA Assays ›
Gene Symbol:
Myl1
Gene Name:
myosin, light chain 1
Gene Aliases:
MLC3-f, MLCf, Mcl3, Mlc1, Mlc3
Chromosome Location:
Chr.9: 73937820 - 73958480 on Build Rnor_6.0
UniGene ID:
Rn.84920
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s132758
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Myl1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077656.1
NP_001071124.1
4
435
NM_020104.2
NP_064489.1
4
253
XM_008767294.1
XP_008765516.1
3
383
XM_008767295.1
XP_008765517.1
3
504
GenBank mRNA
CR755322.1
-
4
258
CR756356.1
-
4
253
FQ214580.1
-
4
562
FQ214608.1
-
4
440
FQ214631.1
-
4
422
FQ214638.1
-
4
529
FQ214656.1
-
4
271
FQ214675.1
-
4
529
FQ214712.1
-
4
443
FQ214723.1
-
4
443
FQ214727.1
-
4
443
FQ214742.1
-
4
443
FQ214939.1
-
4
440
FQ215002.1
-
4
529
FQ215050.1
-
4
443
FQ215083.1
-
4
440
FQ215086.1
-
4
273
FQ215201.1
-
4
440
FQ215229.1
-
4
534
FQ215325.1
-
4
529
FQ215340.1
-
4
271
FQ215418.1
-
4
529
FQ215435.1
-
4
440
FQ215492.1
-
4
422
FQ215500.1
-
3
1019
FQ215535.1
-
3
504
FQ215557.1
-
4
529
FQ215563.1
-
4
440
FQ215578.1
-
4
529
FQ215610.1
-
4
443
FQ215748.1
-
3
502
FQ215766.1
-
4
529
FQ215774.1
-
4
440
FQ215820.1
-
4
498
FQ215848.1
-
4
440
FQ215875.1
-
4
440
FQ215911.1
-
4
534
FQ215933.1
-
4
443
FQ216034.1
-
4
443
FQ216109.1
-
4
440
FQ216124.1
-
4
534
FQ216181.1
-
4
422
FQ216192.1
-
4
529
FQ216213.1
-
5
437
FQ216218.1
-
4
440
FQ216229.1
-
4
710
FQ216238.1
-
4
529
FQ216308.1
-
4
271
FQ216407.1
-
4
611
FQ216452.1
-
4
440
FQ216455.1
-
4
444
FQ216484.1
-
4
422
FQ216496.1
-
4
443
FQ216525.1
-
4
443
FQ216543.1
-
4
440
FQ216556.1
-
4
529
FQ216567.1
-
4
440
FQ216631.1
-
4
443
FQ216654.1
-
4
527
FQ216704.1
-
4
443
FQ216738.1
-
4
271
FQ217074.1
-
4
270
FQ217103.1
-
4
271
FQ217157.1
-
3
246
FQ217192.1
-
4
271
FQ217218.1
-
4
271
FQ217243.1
-
4
270
FQ217303.1
-
4
422
FQ217323.1
-
4
271
FQ217387.1
-
4
271
FQ217394.1
-
4
373
FQ217427.1
-
4
276
FQ217481.1
-
4
271
FQ217524.1
-
4
271
FQ217753.1
-
4
276
FQ217756.1
-
4
271
FQ217889.1
-
4
271
FQ217973.1
-
4
276
FQ217979.1
-
4
271
FQ218029.1
-
4
271
FQ218049.1
-
4
271
FQ223584.1
-
4
529
FQ223632.1
-
4
443
FQ223641.1
-
4
529
FQ223687.1
-
4
271
FQ223755.1
-
4
439
FQ223779.1
-
4
271
FQ223827.1
-
4
443
FQ223880.1
-
4
341
FQ223949.1
-
4
269
FQ223984.1
-
4
271
FQ224047.1
-
4
271
FQ224050.1
-
4
271
FQ224087.1
-
4
271
FQ224112.1
-
3
246
FQ224155.1
-
4
276
FQ224171.1
-
4
440
FQ224172.1
-
4
271
FQ224212.1
-
4
423
FQ224254.1
-
4
271
FQ224277.1
-
4
443
FQ224321.1
-
4
275
FQ224386.1
-
4
276
FQ224411.1
-
4
271
FQ224420.1
-
4
271
FQ224452.1
-
4
346
FQ224519.1
-
4
440
FQ224609.1
-
5
299
FQ224663.1
-
4
271
FQ224726.1
-
4
373
FQ224796.1
-
4
271
FQ224799.1
-
4
276
FQ224927.1
-
4
529
FQ224931.1
-
4
440
FQ224934.1
-
4
438
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_021285, NM_001113387, BC059087, AK003182, AK009938, and others.
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cardiac muscle contraction
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-wh4bk:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0