Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYL1 SIRNA Assays ›
Gene Symbol:
Myl1
Gene Name:
myosin, light chain 1
Gene Aliases:
MLC3-f, MLCf, Mcl3, Mlc1, Mlc3
Chromosome Location:
Chr.9: 73937820 - 73958480 on Build Rnor_6.0
UniGene ID:
Rn.84920
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s132760
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Myl1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077656.1
NP_001071124.1
3
399
NM_020104.2
NP_064489.1
3
217
XM_008767294.1
XP_008765516.1
2
347
XM_008767295.1
XP_008765517.1
2
468
GenBank mRNA
CR755322.1
-
3
222
CR756356.1
-
3
217
FQ214580.1
-
3
526
FQ214604.1
-
3
444
FQ214608.1
-
3
404
FQ214631.1
-
3
386
FQ214638.1
-
3
493
FQ214656.1
-
3
235
FQ214675.1
-
3
493
FQ214712.1
-
3
407
FQ214723.1
-
3
407
FQ214727.1
-
3
407
FQ214742.1
-
3
407
FQ214939.1
-
3
404
FQ215002.1
-
3
493
FQ215050.1
-
3
407
FQ215083.1
-
3
404
FQ215086.1
-
3
237
FQ215201.1
-
3
404
FQ215229.1
-
3
498
FQ215325.1
-
3
493
FQ215340.1
-
3
235
FQ215418.1
-
3
493
FQ215435.1
-
3
404
FQ215492.1
-
3
386
FQ215500.1
-
2
983
FQ215557.1
-
3
493
FQ215563.1
-
3
404
FQ215578.1
-
3
493
FQ215610.1
-
3
407
FQ215748.1
-
2
466
FQ215766.1
-
3
493
FQ215774.1
-
3
404
FQ215820.1
-
3
462
FQ215848.1
-
3
404
FQ215875.1
-
3
404
FQ215911.1
-
3
498
FQ215933.1
-
3
407
FQ216034.1
-
3
407
FQ216109.1
-
3
404
FQ216124.1
-
3
498
FQ216181.1
-
3
386
FQ216192.1
-
3
493
FQ216213.1
-
4
401
FQ216218.1
-
3
404
FQ216229.1
-
3
674
FQ216238.1
-
3
493
FQ216308.1
-
3
235
FQ216407.1
-
3
575
FQ216452.1
-
3
404
FQ216455.1
-
3
408
FQ216484.1
-
3
386
FQ216496.1
-
3
407
FQ216525.1
-
3
407
FQ216543.1
-
3
404
FQ216556.1
-
3
493
FQ216567.1
-
3
404
FQ216631.1
-
3
407
FQ216654.1
-
3
491
FQ216704.1
-
3
407
FQ216737.1
-
3
490
FQ216738.1
-
3
235
FQ217074.1
-
3
234
FQ217103.1
-
3
235
FQ217157.1
-
2
210
FQ217192.1
-
3
235
FQ217218.1
-
3
235
FQ217243.1
-
3
234
FQ217303.1
-
3
386
FQ217323.1
-
3
235
FQ217387.1
-
3
235
FQ217394.1
-
3
337
FQ217427.1
-
3
240
FQ217481.1
-
3
235
FQ217524.1
-
3
235
FQ217753.1
-
3
240
FQ217756.1
-
3
235
FQ217889.1
-
3
235
FQ217973.1
-
3
240
FQ217979.1
-
3
235
FQ218029.1
-
3
235
FQ218049.1
-
3
235
FQ223584.1
-
3
493
FQ223632.1
-
3
407
FQ223641.1
-
3
493
FQ223687.1
-
3
235
FQ223755.1
-
3
403
FQ223779.1
-
3
235
FQ223827.1
-
3
407
FQ223880.1
-
3
305
FQ223949.1
-
3
233
FQ223984.1
-
3
235
FQ224047.1
-
3
235
FQ224050.1
-
3
235
FQ224087.1
-
3
235
FQ224112.1
-
2
210
FQ224155.1
-
3
240
FQ224171.1
-
3
404
FQ224172.1
-
3
235
FQ224254.1
-
3
235
FQ224277.1
-
3
407
FQ224321.1
-
3
239
FQ224386.1
-
3
240
FQ224411.1
-
3
235
FQ224420.1
-
3
235
FQ224452.1
-
3
310
FQ224519.1
-
3
404
FQ224609.1
-
4
263
FQ224663.1
-
3
235
FQ224726.1
-
3
337
FQ224796.1
-
3
235
FQ224799.1
-
3
240
FQ224867.1
-
3
493
FQ224927.1
-
3
493
FQ224931.1
-
3
404
FQ224934.1
-
3
402
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cardiac muscle contraction
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-wh4bk:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0