Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SORL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003105.5 | NP_003096.1 | 24 | 3529 | ||
XM_011542963.2 | XP_011541265.1 | 23 | 3290 | |||
XM_011542965.2 | XP_011541267.1 | 18 | 2438 | |||
XM_011542967.2 | XP_011541269.1 | 4 | 470 | |||
XM_017018169.1 | XP_016873658.1 | 24 | 3236 | |||
XM_017018170.1 | XP_016873659.1 | 22 | 2912 | |||
XM_017018171.1 | XP_016873660.1 | 24 | 3404 | |||
XM_017018172.1 | XP_016873661.1 | 10 | 1401 | |||
GenBank mRNA | AK295878.1 | - | 4 | 470 | ||
BC137171.1 | - | 24 | 3452 | 9021168 | ||
Y08110.1 | - | 24 | 3480 |