Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022186.1 | NP_071522.1 | 4 | 690 | ||
XM_008772913.1 | XP_008771135.1 | 3 | 666 | |||
GenBank mRNA | AB024930.1 | - | 4 | 690 |
Gene Symbol: LOC100363472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728074.4 | XP_002728120.1 | 1 | 638 | ||
XM_008770073.1 | XP_008768295.1 | 2 | 695 |
Gene Symbol: RGD1560263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761340.1 | XP_008759562.1 | 2 | 602 | ||
XM_574989.5 | XP_574989.1 | 1 | 585 |