Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031655.1 | NP_113843.1 | 6 | 790 | ||
GenBank mRNA | BC081763.1 | - | 6 | 776 | 7125782 | |
FQ220594.1 | - | 6 | 805 | |||
U40260.1 | - | 6 | 757 | |||
X76985.1 | - | 6 | 790 |