Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Milr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021585.1 | NP_067596.1 | 3 | 344 | ||
XM_008768388.1 | XP_008766610.1 | 3 | 492 | |||
XM_008768389.1 | XP_008766611.1 | 3 | 492 | |||
GenBank mRNA | U39546.1 | - | 3 | 344 |