Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021659.1 | NP_067691.1 | 9 | 1309 | ||
XM_006231065.2 | XP_006231127.1 | 13 | 2304 | |||
XM_006231066.2 | XP_006231128.1 | 12 | 2700 | |||
XM_006231067.2 | XP_006231129.1 | 12 | 2172 | |||
XM_006231068.2 | XP_006231130.1 | 12 | 2076 | |||
XM_006231069.2 | XP_006231131.1 | 10 | 1584 | |||
GenBank mRNA | AF336852.1 | - | 14 | 2124 | ||
AF336853.1 | - | 13 | 1896 | |||
AF336854.1 | - | 9 | 1149 | |||
AF336855.1 | - | 13 | 2001 | |||
AF336856.1 | - | 11 | 1500 | |||
AF336857.1 | - | 11 | 1509 | |||
AF336858.1 | - | 11 | 1641 | |||
AF336859.1 | - | 12 | 1773 | |||
AF336860.1 | - | 12 | 1869 | |||
U20106.1 | - | 9 | 1309 |