Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017371.4 | NP_001017371.3 | 2 | 1065 | ||
NM_001172712.1 | NP_001166183.1 | 4 | 1407 | |||
NM_003111.4 | NP_003102.1 | 4 | 1416 | |||
GenBank mRNA | AB209334.1 | - | 1 | 529 | ||
AK304199.1 | - | 4 | 988 | |||
AY070137.1 | - | 2 | 1065 | |||
AY441957.1 | - | 4 | 1416 | |||
AY441958.1 | - | 2 | 755 | |||
BC126414.1 | - | 4 | 938 | 8992130 | ||
BC143928.1 | - | 4 | 929 | 9052446 | ||
M97191.2 | - | 2 | 676 | |||
X68560.1 | - | 2 | 634 |