Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021763.1 | NP_068531.1 | 5 | 400 | ||
XM_006232539.2 | XP_006232601.1 | 5 | 661 | |||
XM_006232540.2 | XP_006232602.1 | 6 | 803 | |||
XM_006232541.2 | XP_006232603.1 | 7 | 870 | |||
XM_006232543.1 | XP_006232605.1 | 5 | 413 | |||
XM_006232545.2 | XP_006232607.1 | 4 | 564 | |||
XM_006232546.2 | XP_006232608.1 | 5 | 706 | |||
XM_008761110.1 | XP_008759332.1 | 5 | 417 | |||
XM_008761111.1 | XP_008759333.1 | 5 | 459 | |||
GenBank mRNA | AY004875.1 | - | 5 | 400 | ||
FQ222383.1 | - | 5 | 433 |