Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Syt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031667.2 | NP_113855.2 | 2 | 480 | ||
XM_006232748.1 | XP_006232810.1 | 2 | 472 | |||
GenBank mRNA | AF000423.1 | - | 2 | 521 | ||
AF375465.1 | - | 2 | 279 | |||
BC094518.1 | - | 2 | 480 | 7315033 |