Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Npvf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023952.1 | NP_076442.1 | 2 | 178 | ||
GenBank mRNA | AB040288.1 | - | 2 | 178 | ||
AF330059.2 | - | 2 | 178 |