Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmklr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022218.2 | NP_071554.2 | 2 | 425 | ||
XM_006249505.2 | XP_006249567.1 | 3 | 798 | |||
XM_006249506.2 | XP_006249568.1 | 3 | 477 | |||
XM_006249507.2 | XP_006249569.1 | 3 | 1124 | |||
XM_006249508.2 | XP_006249570.1 | 3 | 741 | |||
XM_006249509.2 | XP_006249571.1 | 3 | 641 | |||
GenBank mRNA | AJ002745.1 | - | 1 | 335 |
Set Membership: |
![]() |