Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hapln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022285.1 | NP_071621.1 | 3 | 254 | ||
XM_006232751.2 | XP_006232813.1 | 3 | 336 | |||
XM_008761208.1 | XP_008759430.1 | 4 | 377 | |||
XM_008761209.1 | XP_008759431.1 | 3 | 562 | |||
GenBank mRNA | AB049056.1 | - | 3 | 254 |