Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022297.2 | NP_071633.1 | 2 | 471 | ||
GenBank mRNA | FQ220049.1 | - | 2 | 491 |