Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4ka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022301.1 | NP_071637.1 | 37 | 4241 | ||
XM_008768902.1 | XP_008767124.1 | 37 | 4394 | |||
GenBank mRNA | D83538.1 | - | 37 | 4519 |