Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001635.3 | NP_001626.1 | 16 | 1443 | ||
NM_139316.2 | NP_647477.1 | 16 | 1443 | |||
XM_006715689.3 | XP_006715752.1 | 16 | 3106 | |||
XM_006715690.3 | XP_006715753.1 | 16 | 1888 | |||
XM_011515271.2 | XP_011513573.1 | 16 | 1888 | |||
XM_017011995.1 | XP_016867484.1 | 16 | 3106 | |||
XM_017011996.1 | XP_016867485.1 | 16 | 2194 | |||
GenBank mRNA | AF034996.1 | - | 16 | 1296 | ||
AF498092.1 | - | 5 | 624 | |||
AF498093.1 | - | 3 | 581 | |||
AF498094.1 | - | 3 | 1493 | |||
AF498095.1 | - | 3 | 275 | |||
AF498096.1 | - | 8 | 1019 | |||
AK291447.1 | - | 16 | 1247 | |||
BC034376.1 | - | 16 | 1234 | 5181100 | ||
U07616.1 | - | 16 | 1337 | |||
X81438.1 | - | 16 | 1301 |