Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEC SIRNA Assays ›
Gene Symbol:
Plec
Gene Name:
plectin
Gene Aliases:
Pcn, Plec1, Pltn
Chromosome Location:
Chr.7: 117230319 - 117291473 on Build Rnor_6.0
UniGene ID:
Rn.1085
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s133909
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Plec
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164296.1
NP_001157768.1
22
2785
NM_001164297.1
NP_001157769.1
22
2720
NM_001164298.1
NP_001157770.1
22
2779
NM_001164299.1
NP_001157771.1
22
2811
NM_001164302.1
NP_001157774.1
22
2765
NM_001164303.1
NP_001157775.1
22
2718
NM_001164304.1
NP_001157776.1
22
2978
NM_001164305.1
NP_001157777.1
22
2695
NM_001164307.1
NP_001157779.1
22
2792
NM_001164308.1
NP_001157780.1
22
2852
NM_022401.2
NP_071796.2
22
3171
XM_006241870.2
XP_006241932.1
23
3382
XM_006241875.2
XP_006241937.1
23
3144
XM_006241882.2
XP_006241944.1
23
3499
XM_008765592.1
XP_008763814.1
23
3975
XM_008765593.1
XP_008763815.1
24
3159
XM_008765594.1
XP_008763816.1
23
4142
XM_008765595.1
XP_008763817.1
23
4946
XM_008765596.1
XP_008763818.1
23
2769
XM_008765597.1
XP_008763819.1
23
2892
XM_008765598.1
XP_008763820.1
23
3382
GenBank mRNA
AJ635267.2
-
5
588
AY480022.1
-
22
2788
AY480023.1
-
22
2723
AY480024.1
-
22
2782
AY480025.1
-
22
2814
AY480026.1
-
22
2768
AY480027.1
-
22
3315
AY480028.1
-
22
2721
AY480029.1
-
22
2866
AY480030.1
-
22
2698
AY480031.1
-
22
2795
AY480032.1
-
22
2855
X59601.1
-
22
3121
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
female pregnancy
response to nutrient
epithelial cell differentiation
hemidesmosome assembly
actin binding
structural constituent of cytoskeleton
cytoskeletal protein binding
structural constituent of muscle
ankyrin binding
poly(A) RNA binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-9ljwx:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0