Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022524.1 | NP_071969.1 | 7 | 901 | ||
XM_006256683.2 | XP_006256745.1 | 6 | 929 | |||
XM_008773077.1 | XP_008771299.1 | 6 | 2055 | |||
GenBank mRNA | BC087639.1 | - | 6 | 900 | 7309360 | |
D78359.1 | - | 7 | 901 |