Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd200r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023953.1 | NP_076443.1 | 5 | 740 | ||
XM_006248347.2 | XP_006248409.1 | 5 | 1053 | |||
XM_008768743.1 | XP_008766965.1 | 5 | 1040 |