Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022529.1 | NP_071974.1 | 5 | 332 | ||
GenBank mRNA | BC059936.1 | - | 5 | 338 | ||
U62635.1 | - | 5 | 332 |