Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Des | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022531.1 | NP_071976.1 | 5 | 1040 | ||
GenBank mRNA | BC061872.1 | - | 5 | 1033 | 5598951 |