Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Araf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033663.2 | NP_001028835.1 | 5 | 549 | ||
NM_022532.2 | NP_071977.1 | 5 | 549 | |||
XM_008773036.1 | XP_008771258.1 | 5 | 1291 | |||
GenBank mRNA | AB158253.1 | - | 4 | 445 | ||
BC060568.1 | - | 589 | ||||
FQ213741.1 | - | 1 | 121 | |||
X06942.1 | - | 4 | 516 |