Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022536.2 | NP_071981.1 | 2 | 225 | ||
GenBank mRNA | AF071225.1 | - | 2 | 225 | ||
BC061971.1 | - | 2 | 254 | 5623342 | ||
FQ211358.1 | - | 2 | 229 | |||
FQ212031.1 | - | 2 | 254 | |||
FQ219792.1 | - | 2 | 263 | |||
FQ219926.1 | - | 2 | 275 | |||
FQ221020.1 | - | 2 | 278 | |||
FQ221248.1 | - | 2 | 273 | |||
FQ221362.1 | - | 2 | 252 | |||
FQ222696.1 | - | 2 | 387 | |||
FQ223162.1 | - | 2 | 272 | |||
FQ228087.1 | - | 2 | 268 | |||
FQ229174.1 | - | 2 | 275 | |||
FQ229214.1 | - | 2 | 269 | |||
FQ230004.1 | - | 279 | ||||
FQ231656.1 | - | 2 | 268 | |||
FQ232077.1 | - | 2 | 264 | |||
FQ232920.1 | - | 2 | 278 |