Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022536.2 | NP_071981.1 | 1, 2 | 134 | ||
GenBank mRNA | AF071225.1 | - | 1, 2 | 134 | ||
BC061971.1 | - | 1, 2 | 163 | 5623342 | ||
FQ211358.1 | - | 1, 2 | 138 | |||
FQ212031.1 | - | 1, 2 | 163 | |||
FQ219792.1 | - | 1, 2 | 172 | |||
FQ219926.1 | - | 1, 2 | 184 | |||
FQ221020.1 | - | 1, 2 | 187 | |||
FQ221248.1 | - | 1, 2 | 182 | |||
FQ221362.1 | - | 1, 2 | 161 | |||
FQ222696.1 | - | 1, 2 | 296 | |||
FQ228087.1 | - | 1, 2 | 177 | |||
FQ229174.1 | - | 1, 2 | 184 | |||
FQ229214.1 | - | 1, 2 | 178 | |||
FQ230004.1 | - | 188 | ||||
FQ231656.1 | - | 1, 2 | 177 | |||
FQ232077.1 | - | 1, 2 | 173 | |||
FQ232920.1 | - | 1, 2 | 187 |