Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022612.1 | NP_072134.1 | 4 | 344 | ||
NM_171988.2 | NP_741985.1 | 4 | 775 | |||
NM_171989.1 | NP_741986.1 | 3 | 254 | |||
XM_006234983.2 | XP_006235045.2 | 4 | 893 | |||
XM_006234985.2 | XP_006235047.2 | 5 | 725 | |||
XM_006234987.2 | XP_006235049.2 | 4 | 635 | |||
GenBank mRNA | AF065431.1 | - | 3 | 203 | ||
AF065432.1 | - | 3 | 254 | |||
AF065433.1 | - | 3 | 512 | |||
AF136927.1 | - | 4 | 344 |