Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022620.1 | NP_072142.1 | 29 | 4098 | ||
XM_008760793.1 | XP_008759015.1 | 32 | 4542 | |||
GenBank mRNA | AF102129.1 | - | 29 | 4098 |
Gene Symbol: LOC100912019 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760836.1 | XP_008759058.1 | 8 | 905 |